missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromobutoxy)phthalimide, 98+%, Thermo Scientific™

Product Code. 11332987
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332987 1 g 1g
11342987 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332987 Supplier Thermo Scientific Alfa Aesar Supplier No. L16314.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5093-32-3
Molecular Formula C12H12BrNO3
Molecular Weight (g/mol) 298.136
MDL Number MFCD00517028
InChI Key HNQARTGCJCNOSX-UHFFFAOYSA-N
Synonym n-4-bromobutoxy phthalimide, 2-4-bromobutoxy isoindoline-1,3-dione, 2-4-bromobutoxy isoindole-1,3-dione, acmc-1arqg, 2-4-bromobutoxy-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutoxy
PubChem CID 7020660
IUPAC Name 2-(4-bromobutoxy)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)OCCCCBr

Specifications

Melting Point 70°C to 72°C
Quantity 1 g
Beilstein 1533519
Formula Weight 298.14
Percent Purity ≥98%
Chemical Name or Material N-(4-Bromobutoxy)phthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.