accessibility menu, dialog, popup

UserName

ONOOBr

CAS RN 5093-32-3

IUPAC Name: 2-(4-bromobutoxy)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(4-bromobutoxy)isoindole-1,3-dione
Molecular Weight (g/mol): 298.14
Molecular Formula: C12H12BrNO3
InChi Key: HNQARTGCJCNOSX-UHFFFAOYSA-N
SMILES: BrCCCCON1C(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results