Learn More
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol, Thermo Scientific™
£23.65 - £265.00
Chemical Identifiers
| CAS | 35141-36-7 |
|---|---|
| Molecular Formula | C9H24ClNO3Si |
| Molecular Weight (g/mol) | 257.83 |
| MDL Number | MFCD00054225 |
| InChI Key | FYZFRYWTMMVDLR-UHFFFAOYSA-M |
| Synonym | n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium |
| PubChem CID | 94462 |
| IUPAC Name | trimethyl(3-trimethoxysilylpropyl)azanium;chloride |
| SMILES | C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15485775
|
Thermo Scientific Alfa Aesar
H66414.14 |
25g |
£26.10 £23.65 / 25g Save £2.45 9% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15495775
|
Thermo Scientific Alfa Aesar
H66414.22 |
100g |
£62.80 £56.65 / 100g Save £6.15 9% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15405785
|
Thermo Scientific Alfa Aesar
H66414.36 |
500g |
£265.00
500g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 35141-36-7 | |
| 257.83 | |
| FYZFRYWTMMVDLR-UHFFFAOYSA-M | |
| 94462 | |
| C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-] |
| C9H24ClNO3Si | |
| MFCD00054225 | |
| n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium | |
| trimethyl(3-trimethoxysilylpropyl)azanium;chloride |
Specifications
| 35141-36-7 | |
| 11°C (52°F) | |
| 1.397 | |
| UN1230 | |
| Hygroscopic | |
| FYZFRYWTMMVDLR-UHFFFAOYSA-M | |
| trimethyl(3-trimethoxysilylpropyl)azanium;chloride | |
| 94462 | |
| N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, in 50% methanol |
| 0.930 g/mL | |
| C9H24ClNO3Si | |
| MFCD00054225 | |
| 11593127 | |
| n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium | |
| C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-] | |
| 257.83 | |
| 257.83 |
Safety and Handling
GHS H Statement
H225-H311-H331-H370-H335-H302-H315-H319
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
missing translation for 'einecsNumber' : 252-393-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- N-Trimethoxysilylpropyl-N,N,N-trimethylammonium chloride, 50% in Methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contain: Methanol, N-Trimethoxysilylpropyl-N,N,N-trimethylammonium chloride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.