missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar H66414.22

Quantity 100g

 View more versions of this product

Product Code. 15495775

  • ONLINE EXCLUSIVE PRICE
    £62.80
    £56.65 / 100g
    Save £6.15
    9% Off
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

35141-36-7
257.83
FYZFRYWTMMVDLR-UHFFFAOYSA-M
94462
C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-]
C9H24ClNO3Si
MFCD00054225
n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium
trimethyl(3-trimethoxysilylpropyl)azanium;chloride

Specifications

35141-36-7
11°C (52°F)
1.397
100g
11593127
n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium
C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-]
257.83
257.83
0.930 g/mL
C9H24ClNO3Si
MFCD00054225
UN1230
Hygroscopic
FYZFRYWTMMVDLR-UHFFFAOYSA-M
trimethyl(3-trimethoxysilylpropyl)azanium;chloride
94462
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, in 50% methanol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.