missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl-L-proline, TRC

Product Code. 30406974
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406974 250 mg 250mg
30390224 100 mg -
30390191 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406974 Supplier Toronto Research Chemicals Supplier No. TRCH296965250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565453-40-9
Molecular Formula C17 H26 N2 O4
Molecular Weight (g/mol) 322.399
Synonym N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl-L-proline, (2S)-1-[2-[(3-Hydroxyadamantan-1-yl)amino]acetyl]pyrrolidine-2-carboxylic Acid
IUPAC Name (2S)-1-[2-[(3-hydroxy-1-adamantyl)amino]acetyl]pyrrolidine-2-carboxylic acid
SMILES OC(=O)[C@@H]1CCCN1C(=O)CNC23CC4CC(CC(O)(C4)C2)C3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H26N2O4/c20-14(19-3-1-2-13(19)15(21)22)9-18-16-5-11-4-12(6-16)8-17(23,7-11)10-16/h11-13,18,23H,1-10H2,(H,21,22)/t11?,12?,13-,16?,17?/m0/s1
Formula Weight 322.189
Percent Purity >95%
Chemical Name or Material (2S)-1-[2-[(3-Hydroxyadamantan-1-yl)amino]acetyl]pyrrolidine-2-carboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.