CAS RN 565453-40-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl-L-proline, TRC
CAS: 565453-40-9 Molecular Formula: C17 H26 N2 O4 Molecular Weight (g/mol): 322.399 Synonym: N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl-L-proline,(2S)-1-[2-[(3-Hydroxyadamantan-1-yl)amino]acetyl]pyrrolidine-2-carboxylic Acid IUPAC Name: (2S)-1-[2-[(3-hydroxy-1-adamantyl)amino]acetyl]pyrrolidine-2-carboxylic acid SMILES: OC(=O)[C@@H]1CCCN1C(=O)CNC23CC4CC(CC(O)(C4)C2)C3