missing translation for 'onlineSavingsMsg'
Learn More

N,3-Diphenyl-2-(2-methyl-1-oxopropyl)4-oxo-N-benzenebutanamide (Mixture of Diastereomers), TRC

Product Code. 30384032
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384032 100 mg 100mg
30384719 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384032 Supplier Toronto Research Chemicals Supplier No. TRCD491820100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444577-70-2
Molecular Formula C26 H25 N O3
Molecular Weight (g/mol) 399.48
Synonym α-(2-Methyl-1-oxopropyl)-γ-oxo-N,β-diphenylbenzenebutanamide
IUPAC Name 4-methyl-3-oxo-2-(2-oxo-1,2-diphenylethyl)-N-phenylpentanamide
SMILES CC(C)C(=O)C(C(C(=O)c1ccccc1)c2ccccc2)C(=O)Nc3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C26H25NO3/c1-18(2)24(28)23(26(30)27-21-16-10-5-11-17-21)22(19-12-6-3-7-13-19)25(29)20-14-8-4-9-15-20/h3-18,22-23H,1-2H3,(H,27,30)
Formula Weight 399.1834
Chemical Name or Material N,3-Diphenyl-2-(2-methyl-1-oxopropyl)4-oxo-N-benzenebutanami
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.