CAS RN 444577-70-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,3-Diphenyl-2-(2-methyl-1-oxopropyl)4-oxo-N-benzenebutanamide (Mixture of Diastereomers), TRC
CAS: 444577-70-2 Molecular Formula: C26 H25 N O3 Molecular Weight (g/mol): 399.48 Synonym: α-(2-Methyl-1-oxopropyl)-γ-oxo-N,β-diphenylbenzenebutanamide IUPAC Name: 4-methyl-3-oxo-2-(2-oxo-1,2-diphenylethyl)-N-phenylpentanamide SMILES: CC(C)C(=O)C(C(C(=O)c1ccccc1)c2ccccc2)C(=O)Nc3ccccc3