missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)glycine (~90%), TRC

Product Code. 30136866
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136866 25 mg 25mg
30004260 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136866 Supplier Toronto Research Chemicals Supplier No. TRCH94217525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5835-28-9
Molecular Formula C4 H9 N O3
Molecular Weight (g/mol) 119.12
Synonym N-(2-Hydroxyethyl)aminoacetic Acid, NSC 15828, Petalonine;
IUPAC Name 2-(2-hydroxyethylamino)acetic acid
SMILES OCCNCC(=O)O

Specifications

InChI Formula InChI=1S/C4H9NO3/c6-2-1-5-3-4(7)8/h5-6H,1-3H2,(H,7,8)
Quantity 25 mg
Formula Weight 119.0582
Shipping Condition Room Temperature
Chemical Name or Material N-(2-Hydroxyethyl)glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.