CAS RN 5835-28-9
CAS RN 5835-28-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-Hydroxyethyl)glycine (~90%), TRC
CAS: 5835-28-9 Molecular Formula: C4 H9 N O3 Molecular Weight (g/mol): 119.12 Synonym: N-(2-Hydroxyethyl)aminoacetic Acid,NSC 15828,Petalonine; IUPAC Name: 2-(2-hydroxyethylamino)acetic acid SMILES: OCCNCC(=O)O