missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)-1-deoxy-L-idonojirimycin Hydrochloride, TRC

Product Code. 30388962
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388962 25 mg 25mg
30404717 2.5 mg 2.5mg
30403323 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388962 Supplier Toronto Research Chemicals Supplier No. TRCH94203025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1322625-34-2
Molecular Formula C8H18ClNO5
Molecular Weight (g/mol) 243.69
Synonym (2S,3R,4R,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)-3,4,5-piperidinetriol Hydrochloride
SMILES O[C@H]1[C@H](O)[C@H](CN(CCO)[C@H]1CO)O.Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H17NO5.ClH/c10-2-1-9-3-6(12)8(14)7(13)5(9)4-11;/h5-8,10-14H,1-4H2;1H/t5-,6-,7+,8+;/m0./s1
Chemical Name or Material N-(2-Hydroxyethyl)-1-deoxy-L-idonojirimycin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.