CAS RN 1322625-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-Hydroxyethyl)-1-deoxy-L-idonojirimycin Hydrochloride, TRC
CAS: 1322625-34-2 Molecular Formula: C8H18ClNO5 Molecular Weight (g/mol): 243.69 Synonym: (2S,3R,4R,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)-3,4,5-piperidinetriol Hydrochloride SMILES: O[C@H]1[C@H](O)[C@H](CN(CCO)[C@H]1CO)O.Cl