missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyano-3-hydroxy-2-propenyl)formamide Monosodium Salt, TRC

Product Code. 30358983
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358983 100 mg 100mg
30354078 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30358983 Supplier Toronto Research Chemicals Supplier No. TRCC981290100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106030-08-4
Molecular Formula C5H5N2NaO2
Molecular Weight (g/mol) 148.1
Synonym Formamide, N-(2-cyano-3-hydroxy-2-propen-1-yl)-, sodium salt (1:1) (ACI), Formamide, N-(2-cyano-3-hydroxy-2-propenyl)-, monosodium salt (9CI), N-(2-Cyano-3-hydroxy-2-propen-1-yl)formamide monosodium salt, Sodium 2-cyano-3-formamidoprop-1-en-1-olate
IUPAC Name sodium;2-cyano-3-formamidoprop-1-en-1-olate
SMILES [Na+].[O-]C=C(CNC=O)C#N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H6N2O2.Na/c6-1-5(3-8)2-7-4-9;/h3-4,8H,2H2,(H,7,9);/q;+1/p-1
Formula Weight 148.0249
Chemical Name or Material N-(2-Cyano-3-hydroxy-2-propenyl)formamide Monosodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.