CAS RN 106030-08-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-Cyano-3-hydroxy-2-propenyl)formamide Monosodium Salt, TRC
CAS: 106030-08-4 Molecular Formula: C5H5N2NaO2 Molecular Weight (g/mol): 148.1 Synonym: Formamide, N-(2-cyano-3-hydroxy-2-propen-1-yl)-, sodium salt (1:1) (ACI),Formamide, N-(2-cyano-3-hydroxy-2-propenyl)-, monosodium salt (9CI),N-(2-Cyano-3-hydroxy-2-propen-1-yl)formamide monosodium salt,Sodium 2-cyano-3-formamidoprop-1-en-1-olate IUPAC Name: sodium;2-cyano-3-formamidoprop-1-en-1-olate SMILES: [Na+].[O-]C=C(CNC=O)C#N