missing translation for 'onlineSavingsMsg'
Learn More

N-(2-chloroethyl)dibenzylamine hydrochloride, 98%, Thermo Scientific™

Product Code. 10626630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10626630 5 g Glass bottle 5g
10301190 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10626630 Supplier Thermo Scientific Acros Supplier No. 436300050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 55-43-6
Molecular Formula C16H19Cl2N
Molecular Weight (g/mol) 296.24
MDL Number MFCD00012518
InChI Key LZXCEBPGNFLHEQ-UHFFFAOYSA-N
Synonym dibenamine hydrochloride, n-2-chloroethyl dibenzylamine hydrochloride, sympatholytin, n,n-dibenzyl-2-chloroethanamine hydrochloride, n,n-dibenzyl-2-chloroethylamine hydrochloride, dibenzylchlorethamine hydrochloride, dibenzylchlorethylamine hydrochloride, n,n-dibenzylaminoethyl chloride hydrochloride, 2-chloroethyldibenzylamine hydrochloride, 2-dibenzylaminoethyl chloride hydrochloride
PubChem CID 5925
SMILES [H+].[Cl-].ClCCN(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 188°C to 194°C
Color White
Elemental Analysis 63.2 to 66.5% C, 4.5 to 4.9% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Merck Index 15,3009
IUPAC Name hydrogen dibenzyl(2-chloroethyl)amine chloride
Formula Weight 296.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-(2-chloroethyl)dibenzylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Chloroethyl)dibenzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.