accessibility menu, dialog, popup

UserName

HClClN

CAS RN 55-43-6

IUPAC Name: hydrogen dibenzyl(2-chloroethyl)amine chloride
Synonyms: hydrogen dibenamine chloride
Molecular Weight (g/mol): 296.24
Molecular Formula: C16H19Cl2N
InChi Key: LZXCEBPGNFLHEQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCCN(CC1=CC=CC=C1)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results