Learn More
Macherey-Nagel™ N 13 aluminum Closure
Product Code. 10630303 Shop All Macherey-Nagel Products
Click to view available options
Color:
Silver
Unit Size:
Pack of 100
Product Code. 10630303
Brand: Macherey-Nagel™ 70257
Only null left
Add to basket
Add to basket
Specifications
Color Silver
Material Aluminum, Butyl Rubber, PTFE
Septum Butyl dark gray / PTFE gray(only centrically laminated with PTFE, typically called Pharma-Fix)
Quantity 100 Pk.
Thickness (Metric) 2 mm
Type Crimp Cap