missing translation for 'onlineSavingsMsg'
Learn More

N-1-Ethyl-1-demethylethyl Torsemide, TRC

Product Code. 30385254
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385254 10 mg
30370830 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385254 Supplier Toronto Research Chemicals Supplier No. TRCE91300010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58155-35-4
Molecular Formula C15 H18 N4 O3 S
Molecular Weight (g/mol) 334.39
Synonym 1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea, Torasemide, Anhydrous Imp. C (EP)
IUPAC Name 1-ethyl-3-[4-(3-methylanilino)pyridin-3-yl]sulfonylurea
SMILES CCNC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H18N4O3S/c1-3-17-15(20)19-23(21,22)14-10-16-8-7-13(14)18-12-6-4-5-11(2)9-12/h4-10H,3H2,1-2H3,(H,16,18)(H2,17,19,20)
Formula Weight 334.11
Chemical Name or Material 1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.