CAS RN 58155-35-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-1-Ethyl-1-demethylethyl Torsemide, TRC
CAS: 58155-35-4 Molecular Formula: C15 H18 N4 O3 S Molecular Weight (g/mol): 334.39 Synonym: 1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea,Torasemide, Anhydrous Imp. C (EP) IUPAC Name: 1-ethyl-3-[4-(3-methylanilino)pyridin-3-yl]sulfonylurea SMILES: CCNC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2
2
1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.