missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYRIP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | MYRIP |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647741
|
Novus Biologicals
NBP2-94564-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666701
|
Novus Biologicals
NBP2-94564-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYRIP Polyclonal antibody specifically detects MYRIP in Human samples. It is validated for Western BlotSpecifications
| MYRIP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 25924 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp586F1018, exophilin-8, FLJ44025, MGC130034, myosin VIIA and Rab interacting protein, Myosin-VIIa- and Rab-interacting protein, MyRIP, rab effector MyRIP, SLAC2-C, SLAC2CMGC130035, Slp homolog lacking C2 domains c, Slp homologue lacking C2 domains, synaptotagmin-like protein homologue lacking C2 domains-c | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 551-770 of human MYRIP (NP_001271354.1). VSLCNISTEVLKVINATEELIAGSTGPWESPQVPPDRQKGMFPRGTDQVRLDEQLTSLEENVYLAAGTVYGLETQLTELEDAARCIHSGTDETHLADLEDQVATAAAQVHHAELQISDIESRISALTIAGLNIAPCVRFTRRRDQKQRTQVQTIDTSRQQRRKLPAPPVKAEKIETSSVTTIKTFNHNFILQGSSTNRTKERKGTTKDLMEPALESAVMY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title