missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYRIP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94564-0.1ml
This item is not returnable.
View return policy
Description
MYRIP Polyclonal antibody specifically detects MYRIP in Human samples. It is validated for Western Blot
Specifications
| MYRIP | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp586F1018, exophilin-8, FLJ44025, MGC130034, myosin VIIA and Rab interacting protein, Myosin-VIIa- and Rab-interacting protein, MyRIP, rab effector MyRIP, SLAC2-C, SLAC2CMGC130035, Slp homolog lacking C2 domains c, Slp homologue lacking C2 domains, synaptotagmin-like protein homologue lacking C2 domains-c | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 551-770 of human MYRIP (NP_001271354.1). VSLCNISTEVLKVINATEELIAGSTGPWESPQVPPDRQKGMFPRGTDQVRLDEQLTSLEENVYLAAGTVYGLETQLTELEDAARCIHSGTDETHLADLEDQVATAAAQVHHAELQISDIESRISALTIAGLNIAPCVRFTRRRDQKQRTQVQTIDTSRQQRRKLPAPPVKAEKIETSSVTTIKTFNHNFILQGSSTNRTKERKGTTKDLMEPALESAVMY | |
| 0.1 mL | |
| Cell Biology, Neuroscience | |
| 25924 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction