missing translation for 'onlineSavingsMsg'
Learn More

Myricetin, TRC

Product Code. 17973385
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973385 25 mg 25mg
17944115 10 mg 10mg
17920885 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973385 Supplier Toronto Research Chemicals Supplier No. TRCM88410025MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-44-2
Molecular Formula C15 H10 O8
Molecular Weight (g/mol) 318.24
Synonym Flavone, 3,3',4',5,5',7-hexahydroxy- (8CI), 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-1-benzopyran-4-one, 3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7,3',4',5'-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one, Cannabiscetin, LDN 0014058, Myricetin, Myricetol, NCBI 5281672, NSC 407290, 4H-1-Benzopyran-4-one, 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
SMILES OC1=C(Oc2cc(O)cc(O)c2C1=O)c3cc(O)c(O)c(O)c3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H10O8/c16-6-3-7(17)11-10(4-6)23-15(14(22)13(11)21)5-1-8(18)12(20)9(19)2-5/h1-4,16-20,22H
Formula Weight 318.0376
Percent Purity >95%
Chemical Name or Material Myricetin (Cannabiscetin)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.