Learn More
Thermo Scientific™ MYP Selective Agar
Description
Enumerate Bacillus cereus in food samples with a selective and differential medium with diagnostic features that rely upon the failure of Bacillus cereus to utilize mannitol and the ability of most strains to produce phospholipase C, resulting in an opaque halo around the colonies. Thermo Scientific™ MYP Selective Agar includes Polymyxin B which inhibits gram-negative bacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | Tested in accordance with CEN ISO/TS 11133-2. According to ISO 7932 |
| Color | Red orange, transparent to slightly opaque |
| Content And Storage | 2° to 12°C, in the dark |
| Format | 90mm Monoplate |
| Manufacturing Origin | Germany |
| Media Type | MYP Selective Agar |
| Product Type | Agar Plate |
| Fill Weight | 17g ± 5% |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.