missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MYP Selective Agar

Product Code. 13215369
Change view
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
MYP Selective Agar
Manufacturing Origin:
Germany
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Media Type Manufacturing Origin Unit Size
13215369 10/Pk. 90mm Monoplate MYP Selective Agar Germany Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13215369 Supplier Thermo Scientific™ Supplier No. PO5133A
  • ONLINE EXCLUSIVE PRICE
    £42.80
    £30.28 / Pack of 10
    Save £12.52
    29% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enumerate Bacillus cereus in food samples while conforming to ISO 7932 with a selective medium proven to be effective with even the most challenging of organism cell ratios.

Enumerate Bacillus cereus in food samples with a selective and differential medium with diagnostic features that rely upon the failure of Bacillus cereus to utilize mannitol and the ability of most strains to produce phospholipase C, resulting in an opaque halo around the colonies. Thermo Scientific™ MYP Selective Agar includes Polymyxin B which inhibits gram-negative bacteria.

Bacillus cereus has been recognized as a causative agent of food poisoning since the 1950s. Many early isolation techniques used blood agar, relying on haemolysis and colony morphology, followed by a range of tests for confirmation. MYP Agar has proven to be very effective for detecting B. cereus even with very low numbers. The formulation conforms the ISO 7932.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested in accordance with CEN ISO/TS 11133-2. According to ISO 7932
Color Red orange, transparent to slightly opaque
Content And Storage 2° to 12°C, in the dark
Format 90mm Monoplate
Manufacturing Origin Germany
Media Type MYP Selective Agar
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.