missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 5a Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | Myosin 5a |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638280
|
Novus Biologicals
NBP2-93397-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668140
|
Novus Biologicals
NBP2-93397-0.1ml |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myosin 5a Polyclonal antibody specifically detects Myosin 5a in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Myosin 5a | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Endocrinology, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4644 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Dilute myosin heavy chain, non-muscle, GS1myosin, heavy polypeptide kinase, MYH12myosin-12, MYO5, Myosin heavy chain 12, myosin V, myosin VA (heavy chain 12, myoxin), myosin VA (heavy polypeptide 12, myoxin), Myosin-12, myosin-Va, Myoxin, MYR12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 950-1150 of human MYO5A (NP_000250.3). LTNLEGIYNSETEKLRSDLERLQLSEEEAKVATGRVLSLQEEIAKLRKDLEQTRSEKKCIEEHADRYKQETEQLVSNLKEENTLLKQEKEALNHRIVQQAKEMTETMEKKLVEETKQLELDLNDERLRYQNLLNEFSRLEERYDDLKEEMTLMVHVPKPGHKRTDSTHSSNESEYIFSSEIAEMEDIPSRTEEPSEKKVPL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title