missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 5a Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93397-0.1ml
This item is not returnable.
View return policy
Description
Myosin 5a Polyclonal antibody specifically detects Myosin 5a in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Myosin 5a | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Dilute myosin heavy chain, non-muscle, GS1myosin, heavy polypeptide kinase, MYH12myosin-12, MYO5, Myosin heavy chain 12, myosin V, myosin VA (heavy chain 12, myoxin), myosin VA (heavy polypeptide 12, myoxin), Myosin-12, myosin-Va, Myoxin, MYR12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 950-1150 of human MYO5A (NP_000250.3). LTNLEGIYNSETEKLRSDLERLQLSEEEAKVATGRVLSLQEEIAKLRKDLEQTRSEKKCIEEHADRYKQETEQLVSNLKEENTLLKQEKEALNHRIVQQAKEMTETMEKKLVEETKQLELDLNDERLRYQNLLNEFSRLEERYDDLKEEMTLMVHVPKPGHKRTDSTHSSNESEYIFSSEIAEMEDIPSRTEEPSEKKVPL | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Endocrinology, Neuroscience, Signal Transduction | |
| 4644 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction