missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 1B Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | Myosin 1B |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231473
|
Novus Biologicals
NBP3-33271-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230838
|
Novus Biologicals
NBP3-33271-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myosin 1B Monoclonal antibody specifically detects Myosin 1B in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Myosin 1B | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 4430 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MMIa, MMI-alpha, MYH-1c, MYO1B variant protein, Myosin I alpha, myosin IB, myosin-I alpha, myosin-Ib, myr1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 916-1136 of Human Myosin 1B (NP_001123630.1).,, Sequence:, RIFHLWRCKKYRDQFTDQQKLIYEEKLEASELFKDKKALYPSSVGQPFQGAYLEINKNPKYKKLKDAIEEKIIIAEVVNKINRANGKSTSRIFLLTNNNLLLADQKSGQIKSEVPLVDVTKVSMSSQNDGFFAVHLKEGSEAASKGDFLFSSDHLIEMATKLYRTTLSQTKQKLNIEISDEFLVQFRQDKVCVKFIQGNQKNGSVPTCKRKNNRLLEVAVP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title