missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 1B Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33271-20ul
This item is not returnable.
View return policy
Description
Myosin 1B Monoclonal antibody specifically detects Myosin 1B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Myosin 1B | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| MMIa, MMI-alpha, MYH-1c, MYO1B variant protein, Myosin I alpha, myosin IB, myosin-I alpha, myosin-Ib, myr1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 916-1136 of Human Myosin 1B (NP_001123630.1).,, Sequence:, RIFHLWRCKKYRDQFTDQQKLIYEEKLEASELFKDKKALYPSSVGQPFQGAYLEINKNPKYKKLKDAIEEKIIIAEVVNKINRANGKSTSRIFLLTNNNLLLADQKSGQIKSEVPLVDVTKVSMSSQNDGFFAVHLKEGSEAASKGDFLFSSDHLIEMATKLYRTTLSQTKQKLNIEISDEFLVQFRQDKVCVKFIQGNQKNGSVPTCKRKNNRLLEVAVP | |
| 20 μL | |
| Vision | |
| 4430 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction