missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myomegalin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Myomegalin |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230048
|
Novus Biologicals
NBP3-38462-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230956
|
Novus Biologicals
NBP3-38462-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myomegalin Polyclonal antibody specifically detects Myomegalin in Mouse samples. It is validated for ELISA,Western BlotSpecifications
| Myomegalin | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Cardiomyopathy-associated protein 2, CMYA2, DKFZp781J054, KIAA0454, KIAA0477, MGC75440, MMGLcardiomyopathy associated 2, myomegalin, phosphodiesterase 4D interacting protein, Phosphodiesterase 4D-interacting protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human Myomegalin (NP_071754.3).,, Sequence:, MKGTDSGSCCRRRCDFGCCCRASRRAHYTPYRSGDATRTPQSPRQTPSRERRRPEPAGSWAAAAEEEEAAAAATPWMRDYFAEDDGEMVPRTSHTAAFLSDTKDRGPPVQSQIWRSGEKVPFVQTYSLRAFEKPPQVQTQALRDFEKHLNDLKKENFSLKLRIYFLEERMQQKYEASREDIYKRNIELKVEVESLKRELQDKKQHLDKTWADVENLNSQNEAELRRQFEERQQETEHVYELLENKIQLLQEESRLAKNEAARMAALVEAEKECNLELSEKLKGVTKNWEDVPGDQVKPDQYTEALAQRDK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 9659 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title