missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myomegalin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38462-20ul
This item is not returnable.
View return policy
Description
Myomegalin Polyclonal antibody specifically detects Myomegalin in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Myomegalin | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Cardiomyopathy-associated protein 2, CMYA2, DKFZp781J054, KIAA0454, KIAA0477, MGC75440, MMGLcardiomyopathy associated 2, myomegalin, phosphodiesterase 4D interacting protein, Phosphodiesterase 4D-interacting protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human Myomegalin (NP_071754.3).,, Sequence:, MKGTDSGSCCRRRCDFGCCCRASRRAHYTPYRSGDATRTPQSPRQTPSRERRRPEPAGSWAAAAEEEEAAAAATPWMRDYFAEDDGEMVPRTSHTAAFLSDTKDRGPPVQSQIWRSGEKVPFVQTYSLRAFEKPPQVQTQALRDFEKHLNDLKKENFSLKLRIYFLEERMQQKYEASREDIYKRNIELKVEVESLKRELQDKKQHLDKTWADVENLNSQNEAELRRQFEERQQETEHVYELLENKIQLLQEESRLAKNEAARMAALVEAEKECNLELSEKLKGVTKNWEDVPGDQVKPDQYTEALAQRDK | |
| 20 μL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9659 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction