missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myogenin Mouse, Clone: SPM144, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32987-0.02MG
This item is not returnable.
View return policy
Description
Myogenin Monoclonal specifically detects Myogenin in Human, Mouse, Rat, Porcine, Feline samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Myogenin | |
| Monoclonal | |
| 0.2mg/mL | |
| Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
| P15173 | |
| MYOG | |
| Human myogenin recombinant protein | |
| Protein A purified | |
| RUO | |
| Primary | |
| Myogenin is a member of the MyoD family of myogenic basic helix-loop-helix (bHLH) transcription factors that also includes MyoD, Myf-5, and MRF4 (also known as herculinor Myf-6). MyoD family members are expressed exclusively in skeletal muscle and play a key role in activating myogenesis by binding to enhancer sequences of muscle-specific genes. The regulatory domain of MyoD is approximately 70 amino acids in length and includes both a basic DNA binding motif and a bHLH dimerization motif. MyoD family members share about 80% amino acid homology in their bHLH motifs. Anti-myogenin labels the nuclei of myoblasts in developing muscle tissue, and is expressed in tumor cell nuclei of rhabdomyosarcoma and some leiomyosarcomas. Positive nuclear staining may occur in Wilms tumor. | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SPM144 | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
| Mouse | |
| 34 kDa | |
| 0.02 mg | |
| Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
| 4656 | |
| Human, Mouse, Rat, Pig, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction