missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myocilin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4B1F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60357-1-IG-150UL
This item is not returnable.
View return policy
Description
Myocilin is an extracellular protein expressed in the eye, including the retina, trabecular meshwork and ciliary body. Myocilin can form homomultimers in vivo and can also associate with components of the ECM via interactions with the Hep II domain of FibroPVRL1. In addition, myocilin interacts with myosin regulatory light chain, a component of the myosin motor protein complex. This interaction implies a role for myocilin in the actomyosin system, linking myocilin to the functional status of the trabecular meshwork (TM), which is responsible for controlling the intraocular pressure (IOP). Alterations in functions of the TM may lead to IOP elevation and development of glaucoma, a major cause of blindness. Myocilin is encoded by MYOC (also designated TIGR), a gene that maps to the GLC1A locus on chromosome 1q24.3 and is susceptible to mutations. Mutations in the MYOC gene are specifically linked with primary open angle glaucoma (POAG), a blinding disease characterized by progressive loss of retinal ganglion cells.Specifications
| Myocilin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70624, Q99972, Q9R1J4 | |
| Myoc | |
| Myocilin Fusion Protein Ag5492 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4B1F6 | |
| Unconjugated | |
| MYOC | |
| GLC1A, GPOA, JOAG, JOAG1, MYOC, myocilin, TIGR | |
| Mouse | |
| Protein A | |
| RUO | |
| 17926, 397046, 4653, 81523 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction