missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO3A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£385.00 - £587.00
Specifications
| Antigen | MYO3A |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18339023
|
Bio-Techne
NBP3-17742-25UL |
25 μg |
£385.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18305266
|
Novus Biologicals
NBP3-17742-100UL |
100 μg |
£587.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYO3A Polyclonal antibody specifically detects MYO3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MYO3A | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase, Vision | |
| PBS, pH 7.2, 40% glycerol | |
| 53904 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| deafness, autosomal recessive 30, DFNB30, EC 2.7.11, EC 2.7.11.1, myosin IIIA, myosin-IIIa | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KRRPRKDSQGKLLDLEDFYYKEFLPSRSGPKEHSPSLRERRPQQELQNQCIKANERCWAAESPEKEEEREPAANPYDFR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title