missing translation for 'onlineSavingsMsg'
Learn More

MYH6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £375.00

Specifications

Antigen MYH6
Concentration 1000 μg/mL
Applications Immunofluorescence
Conjugate CoraLite Plus 488
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18020726
View Documents
Proteintech
CL488-22281-100UL
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18019375
View Documents
Proteintech
CL488-22281-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MYH6 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH6. This antibody recognizes human, mouse, rat antigen. The MYH6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.

Specifications

Specifications

MYH6
Immunofluorescence
Liquid
Human, Mouse, Rat
alpha MHC, ASD3, MYH6, MYHC, MyHC alpha, MYHCA, Myosin 6, Myosin heavy chain 6
Primary
1000 μg/mL
CoraLite Plus 488
Rabbit
4624
IgG
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.