Learn More
MYH6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-22281-20UL
 View more versions of this product
Product Code. 18019375
  • £135.00 / 20µL
Description
The MYH6 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH6. This antibody recognizes human, mouse, rat antigen. The MYH6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
MYH6
1000 μg/mL
alpha MHC, ASD3, MYH6, MYHC, MyHC alpha, MYHCA, Myosin 6, Myosin heavy chain 6
20 μL
4624
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa