missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £369.00
Specifications
| Antigen | MYH4 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607588
|
Novus Biologicals
NBP3-05634-100ul |
100 μg |
£369.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669477
|
Novus Biologicals
NBP3-05634-20ul |
20 μg |
£159.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYH4 Polyclonal antibody specifically detects MYH4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| MYH4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 4622 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MyHC-2B, MyHC-IIb, myosin heavy chain 4, myosin heavy chain IIb, myosin heavy chain, skeletal muscle, fetal, myosin, heavy chain 4, skeletal muscle, myosin, heavy polypeptide 4, skeletal muscle, myosin-4 | |
| A synthetic peptide corresponding to a sequence within amino acids 1500-1600 of human MYH4 (NP_060003.2). ETLKRENKNLQQEISDLTEQIAEGGKHIHELEKVKKQLDHEKSELQTSLEEAEASLEHEEGKILRIQLELNQVKSEIDRKIAEKDEELDQLKRNHLRVVES | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title