Learn More
MYH4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-05634-100ul
 View more versions of this product
Product Code. 18607588
  • £369.00 / 100µL
Description
MYH4 Polyclonal antibody specifically detects MYH4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MYH4
Polyclonal
Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
MyHC-2B, MyHC-IIb, myosin heavy chain 4, myosin heavy chain IIb, myosin heavy chain, skeletal muscle, fetal, myosin, heavy chain 4, skeletal muscle, myosin, heavy polypeptide 4, skeletal muscle, myosin-4
A synthetic peptide corresponding to a sequence within amino acids 1500-1600 of human MYH4 (NP_060003.2). ETLKRENKNLQQEISDLTEQIAEGGKHIHELEKVKKQLDHEKSELQTSLEEAEASLEHEEGKILRIQLELNQVKSEIDRKIAEKDEELDQLKRNHLRVVES
100 μg
Cell Biology, Cytoskeleton Markers, Signal Transduction
4622
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified