missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH14 Mouse anti-Human, Mouse, Clone: 1B4C9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MYH14 |
|---|---|
| Clone | 1B4C9 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16889055
|
Proteintech
66825-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16879055
|
Proteintech
66825-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.Specifications
| MYH14 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DFNA4, KIAA2034, MHC16, MYH14, myosin, Myosin 14, Myosin heavy chain 14, myosin, heavy chain 14, NMHC II C | |
| Myh14 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1B4C9 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q6URW6, Q7Z406 | |
| 71960, 79784 | |
| MYH14 Fusion Protein Ag28063 | |
| Primary | |
| -20°C | |
| MYH14 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title