missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH14 Mouse anti-Human, Mouse, Clone: 1B4C9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66825-1-IG-150UL
This item is not returnable.
View return policy
Description
Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.Specifications
| MYH14 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6URW6, Q7Z406 | |
| Myh14 | |
| MYH14 Fusion Protein Ag28063 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B4C9 | |
| Unconjugated | |
| MYH14 | |
| DFNA4, KIAA2034, MHC16, MYH14, myosin, Myosin 14, Myosin heavy chain 14, myosin, heavy chain 14, NMHC II C | |
| Mouse | |
| Protein A | |
| RUO | |
| 71960, 79784 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction