missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£173.00 - £310.00
Specifications
| Antigen | MYH |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 1:10- 1:2000. Use reported in scientific literature, Immunocytochemistry/Immunofluorescence 18 ug/ml. Use reported in scientific literature |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18264672
|
Novus Biologicals
NB600-1032SS |
0.025 mg |
£173.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18247742
|
Novus Biologicals
NB600-1032 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYH Polyclonal specifically detects MYH in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| MYH | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:10- 1:2000. Use reported in scientific literature, Immunocytochemistry/Immunofluorescence 18 ug/ml. Use reported in scientific literature | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bacteria | |
| NP_001121897 | |
| 4595 | |
| This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 531-546 of human MYH. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Base Excision Repair, Cancer, DNA Repair | |
| PBS containing 0.05% BSA with 0.02% Sodium Azide | |
| CYP2C, EC 3.2.2, EC 3.2.2.-, hMYH, MGC4416, mutY (E. coli) homolog, MutY homolog, mutY homolog (E. coli), Muyth, MYHA/G-specific adenine DNA glycosylase | |
| MUTYH | |
| IgG | |
| Protein G purified | |
| Cross-reacts with Human, Mouse, Rat and Bacteria. Not yet tested in other species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title