missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB600-1032
This item is not returnable.
View return policy
Description
MYH Polyclonal specifically detects MYH in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| MYH | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.05% BSA with 0.02% Sodium Azide | |
| CYP2C, EC 3.2.2, EC 3.2.2.-, hMYH, MGC4416, mutY (E. coli) homolog, MutY homolog, mutY homolog (E. coli), Muyth, MYHA/G-specific adenine DNA glycosylase | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross-reacts with Human, Mouse, Rat and Bacteria. Not yet tested in other species. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:10- 1:2000. Use reported in scientific literature, Immunocytochemistry/Immunofluorescence 18 ug/ml. Use reported in scientific literature | |
| NP_001121897 | |
| MUTYH | |
| This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 531-546 of human MYH. | |
| 0.1 mg | |
| Base Excision Repair, Cancer, DNA Repair | |
| 4595 | |
| Human, Mouse, Rat, Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction