missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1G Antibody (S178A-9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59322-25ug
This item is not returnable.
View return policy
Description
CACNA1G Monoclonal specifically detects CACNA1G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| CACNA1G | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| CACNA1G | |
| Fusion protein amino acids 2052-2172 (cytoplasmic C-terminus) of mouse Cav3.1 | |
| 25 μg | |
| Primary | |
| Detects <200kDa. Does not cross-react with Cav3.2. | |
| Store at -20°C. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| S178A-9 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 8913 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction