missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1G Antibody (S178A-9), Novus Biologicals™
Mouse Monoclonal Antibody
£180.00 - £440.00
Specifications
| Antigen | CACNA1G |
|---|---|
| Clone | S178A-9 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18257221
|
Novus Biologicals
NBP2-59322 |
100 μg |
£440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633027
|
Novus Biologicals
NBP2-59322-25ug |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNA1G Monoclonal specifically detects CACNA1G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| CACNA1G | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 8913 | |
| Fusion protein amino acids 2052-2172 (cytoplasmic C-terminus) of mouse Cav3.1 | |
| Primary | |
| Detects <200kDa. Does not cross-react with Cav3.2. |
| S178A-9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit | |
| CACNA1G | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title