missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Myeloperoxidase/MPO Antibody (CL15002), Novus Biologicals™
Mouse Monoclonal Antibody
£246.00 - £484.00
Specifications
| Antigen | Myeloperoxidase/MPO |
|---|---|
| Clone | CL15002 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043300
|
Novus Biologicals™
NBP3-26847-25ul |
25 μL |
£246.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043378
|
Novus Biologicals™
NBP3-26847-100ul |
100 μL |
£484.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| Myeloperoxidase/MPO | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| PBS, pH 7.2, 40% glycerol | |
| 4353 | |
| IgG1 | |
| Protein A purified |
| CL15002 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| This antibody was developed against a Synthetic Peptide of P05164. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title