missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Myeloperoxidase/MPO Antibody (CL15002), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26847-100ul

 View more versions of this product

Product Code. 30043378

  • £484.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Myeloperoxidase/MPO
Monoclonal
1 mg/mL
Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500
EC 1.11.1, EC 1.11.1.7, myeloperoxidase
This antibody was developed against a Synthetic Peptide of P05164. Actual immunogen sequence is proprietary.
100 μL
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
4353
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Immunohistochemistry, Western Blot
CL15002
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.