missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Myeloperoxidase/MPO Activity Assay Kit (Fluorometric)

Product Code. 18678610 Shop All Bio Techne Products
Click to view available options
Quantity:
96 Tests
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18678610

Brand: Novus Biologicals™ NBP324524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Assay Kit (Fluorometric)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage of components varies. See protocol for specific instructions.
Gene ID (Entrez) 4353
Gene Symbol MPO
Quantity 96 Tests
Synonym EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Research Category Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Species Non-species specific
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.