Learn More
Myelin Basic Protein Ab-1 Rabbit Polyclonal Antibody, Epredia™
Myelin Basic Protein Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
£166.24 - £833.00
Specifications
| Antigen | Myelin Basic Protein Ab-1 |
|---|---|
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12692917
|
Epredia
RB-1460-A0 |
100 μL | Concentrated | ||||||
|
12602927
|
Epredia
RB-1460-A1 |
500 μL | Concentrated | ||||||
|
12682917
|
Epredia
RB-1460-A |
1 mL | Concentrated | ||||||
|
12632927
|
Epredia
RB-1460-R7 |
7 mL | Prediluted | ||||||
Description
Myelin basic protein accounts for about 30% of the proteins in myelin from the central nervous system. It is best known for its capacity to induce experimental allergic encephalomyelitis (EAE), a T-lymphocyte mediated disease due to delayed-type hypersensitivity; though each animal species appears to respond to a different fragment of the 170 amino acid polypeptide. Myelin basic protein is expressed in oligodendrocytes, myelin of white matter in the brain and spinal cord and in peripheral nerves.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Immunogen: Myelin basic protein isolated from human brain
Positive Control: Brain
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin )
Specifications
| Myelin Basic Protein Ab-1 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| Myelin basic protein isolated from human brain | |
| Primary |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.