missing translation for 'onlineSavingsMsg'
Learn More

Myelin Basic Protein Ab-1 Rabbit Polyclonal Antibody, Epredia™

Myelin Basic Protein Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Brand:  Epredia RB-1460-A

 View more versions of this product

Product Code. 12682917

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Myelin basic protein accounts for about 30% of the proteins in myelin from the central nervous system. It is best known for its capacity to induce experimental allergic encephalomyelitis (EAE), a T-lymphocyte mediated disease due to delayed-type hypersensitivity; though each animal species appears to respond to a different fragment of the 170 amino acid polypeptide. Myelin basic protein is expressed in oligodendrocytes, myelin of white matter in the brain and spinal cord and in peripheral nerves.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: Myelin basic protein isolated from human brain

Positive Control: Brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin )
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Myelin Basic Protein Ab-1
Polyclonal
Rabbit
1 mL
Neuroscience
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
Myelin basic protein isolated from human brain
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.