missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£167.00 - £312.00
Specifications
| Antigen | MyD88 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 20228254) |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18159391
|
Novus Biologicals
NBP2-24671SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18140262
|
Novus Biologicals
NBP2-24671 |
0.1 mg |
£312.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MyD88 Polyclonal specifically detects MyD88 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MyD88 | |
| Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 20228254) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| MYD88 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in horse, opossum, chimpanzee, dog, 88% homologous in rat, mouse and 77% homologous in cow. |
| 0.5 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 4615 | |
| A portion of amino acids 50-100 of human MyD88 was used as immunogen for this antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title