missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-24671
This item is not returnable.
View return policy
Description
MyD88 Polyclonal specifically detects MyD88 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| MyD88 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| MYD88 | |
| A portion of amino acids 50-100 of human MyD88 was used as immunogen for this antibody. | |
| 0.1 mg | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 4615 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 20228254) | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in horse, opossum, chimpanzee, dog, 88% homologous in rat, mouse and 77% homologous in cow. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction